PE-Labeled Human SIRP alpha, Fc Tag (recommended for neutralizing assay) (Cat. No. SIA-HP252) is produced via conjugation of PE to H uman SIRP alpha, Fc tag under optimal conditions. Human SIRP alpha, Fc tag is expressed in HEK293 cells containing AA 1 * 50 Tests
Fournisseur: ACROBIOSYSTEMS
PE-Labeled Human SIRP alpha, Fc Tag (recommended for neutralizing assay) (Cat. No. SIA-HP252) is produced via conjugation of PE to H uman SIRP alpha, Fc tag under optimal conditions. Human SIRP alpha, Fc tag is expressed in HEK293 cells containing AA 1 * 50 Tests
COMMANDE
Développer tout / Réduire tout
PE-Labeled Human SIRP alpha, Fc Tag (recommended for neutralizing assay) (Cat. No. SIA-HP252) is produced via conjugation of PE to H uman SIRP alpha, Fc tag under optimal conditions. Human SIRP alpha, Fc tag is expressed in HEK293 cells containing AA 1 * 50 Tests ACRBSIAHP25250TEST 1 * 50 Tests
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou