Recombinat Protein,Human SOD1 Cu-Zn SOD Protein, His Tag,Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contain s AA Ala 2 - Gln 154 (Accession AAH01034).This protein carries a polyhistidine tag at the N-terminus. The protein has 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human SOD1 Cu-Zn SOD Protein, His Tag,Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contain s AA Ala 2 - Gln 154 (Accession AAH01034).This protein carries a polyhistidine tag at the N-terminus. The protein has 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human SOD1 Cu-Zn SOD Protein, His Tag,Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contain s AA Ala 2 - Gln 154 (Accession AAH01034).This protein carries a polyhistidine tag at the N-terminus. The protein has 1 * 1 mg ACRBSO1-H5148-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou