MABSol Biotinylated Human SOST, His Tag, primary amine labeling (SOT-H8245) is expressed from human HEK293 cells. It contains AA Gl n 24 - Tyr 213 (Accession AAI01087.1). It is the biotinylated form of Human SOST, His Tag (HST-H5245). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Human SOST, His Tag, primary amine labeling (SOT-H8245) is expressed from human HEK293 cells. It contains AA Gl n 24 - Tyr 213 (Accession AAI01087.1). It is the biotinylated form of Human SOST, His Tag (HST-H5245). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human SOST, His Tag, primary amine labeling (SOT-H8245) is expressed from human HEK293 cells. It contains AA Gl n 24 - Tyr 213 (Accession AAI01087.1). It is the biotinylated form of Human SOST, His Tag (HST-H5245). 1 * 25 µG ACRBSOT-H8245-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou