SARS-CoV-2 Spike NTD His Tag (B.1.1.529/Omicron) (SPD-C522d) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Le u 303 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE)). The spike mu entifi 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
SARS-CoV-2 Spike NTD His Tag (B.1.1.529/Omicron) (SPD-C522d) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Le u 303 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE)). The spike mu entifi 1 * 100 G
COMMANDE
Développer tout / Réduire tout
SARS-CoV-2 Spike NTD His Tag (B.1.1.529/Omicron) (SPD-C522d) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Le u 303 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE)). The spike mu entifi 1 * 100 µG ACRBSPDC522D-100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou