Recombinat Protein,Human Stathmin 1 STMN1 Protein, His Tag,Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession NP981946).This protein carries a polyhistidine tag at the N-terminus. The pro c 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Stathmin 1 STMN1 Protein, His Tag,Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession NP981946).This protein carries a polyhistidine tag at the N-terminus. The pro c 1 * 100 µG ACRBST1-H5149100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou