Recombinat Protein,Human TFPI-2 Protein, His Tag,Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It co ntains AA Asp 23 - Lys 213 (Accession NP006519.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human TFPI-2 Protein, His Tag,Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It co ntains AA Asp 23 - Lys 213 (Accession NP006519.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human TFPI-2 Protein, His Tag,Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It co ntains AA Asp 23 - Lys 213 (Accession NP006519.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 50 µG ACRBTF2-H5227-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou