Recombinat Protein,Human TFPI Protein, His Tag,Human TFPI, His Tag (TFI-H5226) is expressed from human 293 cells (HEK293). It contai ns AA Asp 29 - Lys 282 (Accession NP006278.1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human TFPI Protein, His Tag,Human TFPI, His Tag (TFI-H5226) is expressed from human 293 cells (HEK293). It contai ns AA Asp 29 - Lys 282 (Accession NP006278.1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human TFPI Protein, His Tag,Human TFPI, His Tag (TFI-H5226) is expressed from human 293 cells (HEK293). It contai ns AA Asp 29 - Lys 282 (Accession NP006278.1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBTFI-H5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou