MABSol® Biotinylated Human TFPI, His Tag, primary amine labeling (TFI-H8226) is expressed from human HEK293 cells. It contains AA A sp 29 - Lys 282 (Accession NP006278.1). It is the biotinylated form of Human TFPI, His Tag (TFI-H5226). null
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human TFPI, His Tag, primary amine labeling (TFI-H8226) is expressed from human HEK293 cells. It contains AA A sp 29 - Lys 282 (Accession NP006278.1). It is the biotinylated form of Human TFPI, His Tag (TFI-H5226). null ACRBTFI-H8226-25UG Each
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou