Cynomolgus Transferrin R, His Tag (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession XP005545315.1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Transferrin R, His Tag (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession XP005545315.1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus Transferrin R, His Tag (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession XP005545315.1). 1 * 1 mg ACRBTFR-C524A-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou