Recombinat Protein,Human Thrombospondin-2 Protein, His Tag,Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 c ells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession P35442-1).This protein carries a polyhistidine tag at the 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Thrombospondin-2 Protein, His Tag,Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 c ells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession P35442-1).This protein carries a polyhistidine tag at the 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Thrombospondin-2 Protein, His Tag,Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 c ells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession P35442-1).This protein carries a polyhistidine tag at the 1 * 1 mg ACRBTH2-H52H5-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou