Recombinat Protein,Human TIMP1 Protein, His Tag,Human TIMP1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Cys 24 - Ala 207 (Accession AAH00866).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human TIMP1 Protein, His Tag,Human TIMP1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Cys 24 - Ala 207 (Accession AAH00866).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human TIMP1 Protein, His Tag,Human TIMP1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Cys 24 - Ala 207 (Accession AAH00866).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBTI1-H5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou