Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, His Tag,Mouse TIM-3, His Tag (TI3-M52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a polyhistidine tag at the C-terminus. s a 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, His Tag,Mouse TIM-3, His Tag (TI3-M52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a polyhistidine tag at the C-terminus. s a 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, His Tag,Mouse TIM-3, His Tag (TI3-M52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a polyhistidine tag at the C-terminus. s a 1 * 100 µG ACRBTI3-M52H1100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou