Cynomolgus Rhesus macaque TIGIT, His Tag (HPLC-verified) (TIT-C5223) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession XP015300911.1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus ma e 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque TIGIT, His Tag (HPLC-verified) (TIT-C5223) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession XP015300911.1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus ma e 1 * 50 µG ACRBTIT-C5223-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou