Cynomolgus Rhesus macaque TIGIT, Fc Tag (TIT-C5254) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Ac cession G7NXM4-1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homolog 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque TIGIT, Fc Tag (TIT-C5254) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Ac cession G7NXM4-1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homolog 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque TIGIT, Fc Tag (TIT-C5254) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Ac cession G7NXM4-1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homolog 1 * 1 mg ACRBTIT-C5254-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou