Human TIM-3, His Tag (TM3-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession Q8TDQ0-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human TIM-3, His Tag (TM3-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession Q8TDQ0-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human TIM-3, His Tag (TM3-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession Q8TDQ0-1). 1 * 1 mg ACRBTM3-H5229-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou