Human TNFR1, His Tag (TN1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human TNFR1, His Tag (TN1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human TNFR1, His Tag (TN1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 1 mg ACRBTN1-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou