Recombinat Protein,Human DR6 TNFRSF21 Protein, Fc Tag,Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Leu 350 (Accession AAH17730).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human DR6 TNFRSF21 Protein, Fc Tag,Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Leu 350 (Accession AAH17730).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human DR6 TNFRSF21 Protein, Fc Tag,Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Leu 350 (Accession AAH17730).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 1 mg ACRBTN1-H5252-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou