ActiveMax Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211) is expressed from human 293 cells (H EK293). It contains AA Val 77 - Leu 233 (Accession P01375-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
ActiveMax Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211) is expressed from human 293 cells (H EK293). It contains AA Val 77 - Leu 233 (Accession P01375-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211) is expressed from human 293 cells (H EK293). It contains AA Val 77 - Leu 233 (Accession P01375-1). 1 * 100 µG ACRBTNA-H4211100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou