MABSol® Biotinylated Human TNF-alpha, epitope tag free, primary amine labeling (active trimer) (MALS verified) (TNA-H8211) is expres sed from human HEK293 cells. It contains AA Val 77 - Leu 233 (Accession NP000585.2). It is the biotinylated form of n 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human TNF-alpha, epitope tag free, primary amine labeling (active trimer) (MALS verified) (TNA-H8211) is expres sed from human HEK293 cells. It contains AA Val 77 - Leu 233 (Accession NP000585.2). It is the biotinylated form of n 1 * 200 µG ACRBTNA-H8211200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou