MABSol Biotinylated Mouse TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-M82E9) is expressed from human HEK293 cells. I t contains AA Leu 80 - Leu 235 (Accession P06804). 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Mouse TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-M82E9) is expressed from human HEK293 cells. I t contains AA Leu 80 - Leu 235 (Accession P06804). 1 * 200 G
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Mouse TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-M82E9) is expressed from human HEK293 cells. I t contains AA Leu 80 - Leu 235 (Accession P06804). 1 * 200 µG ACRBTNA-M82E9200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou