Human Osteoprotegerin, His Tag (TNB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Leu 401 (Accession N P002537). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human Osteoprotegerin, His Tag (TNB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Leu 401 (Accession N P002537). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human Osteoprotegerin, His Tag (TNB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Leu 401 (Accession N P002537). 1 * 100 µG ACRBTNB-H5220100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou