Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession O95407-1). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession O95407-1). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession O95407-1). 1 * 50 µG ACRBTNB-H5255-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou