Recombinat Protein,Unconjugated Human TPBG 5T4 Protein, His,Avitag,Unconjugated Human TPBG, His,Avitag (TPG-H52E5) is expressed fr om human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidin 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Unconjugated Human TPBG 5T4 Protein, His,Avitag,Unconjugated Human TPBG, His,Avitag (TPG-H52E5) is expressed fr om human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidin 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Unconjugated Human TPBG 5T4 Protein, His,Avitag™,Unconjugated Human TPBG, His,Avitag (TPG-H52E5) is expressed fr om human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidin 1 * 1 mg ACRBTPG-H52E5-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou