Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835 ). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835 ). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835 ). 1 * 1 mg ACRBTR1-H5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou