Human TRAIL R2, His Tag (TR2-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession NP003833 ). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human TRAIL R2, His Tag (TR2-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession NP003833 ). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human TRAIL R2, His Tag (TR2-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession NP003833 ). 1 * 100 µG ACRBTR2-H5229100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou