Recombinat Protein,Human TRAIL R4 TNFRSF10D Protein, His Tag,Human TRAIL R4, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession NP003831).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human TRAIL R4 TNFRSF10D Protein, His Tag,Human TRAIL R4, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession NP003831).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg ACRBTR4-H52H5-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou