Recombinat Protein,Human TRAIL R4 TNFRSF10D Protein, His Tag,Human TRAIL R4, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession NP003831).This protein carries a polyhistidine tag at the C-termi 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human TRAIL R4 TNFRSF10D Protein, His Tag,Human TRAIL R4, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession NP003831).This protein carries a polyhistidine tag at the C-termi 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human TRAIL R4 TNFRSF10D Protein, His Tag,Human TRAIL R4, His Tag (TR4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 56 - His 211 (Accession NP003831).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG ACRBTR4-H52H5100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou