Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (TRN-M525b) is expressed from human 293 cells (HEK293). It contains AA Val 20 - His 697 (Accession Q921I1-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (TRN-M525b) is expressed from human 293 cells (HEK293). It contains AA Val 20 - His 697 (Accession Q921I1-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (TRN-M525b) is expressed from human 293 cells (HEK293). It contains AA Val 20 - His 697 (Accession Q921I1-1). 1 * 100 µG ACRBTRN-M525B100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou