Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein c 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein c 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein c 1 * 100 µG ACRBTSP-C52H4100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou