Human TSLP (R127A, R130A), His Tag (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accessio n Q969D9-1 (R127A, R130A)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human TSLP (R127A, R130A), His Tag (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accessio n Q969D9-1 (R127A, R130A)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human TSLP (R127A, R130A), His Tag (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accessio n Q969D9-1 (R127A, R130A)). 1 * 1 mg ACRBTSP-H52HA-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou