Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 1 mg ACRBTSP-H52HB-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou