Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human TSLP, His Tag (TSP-H52Hb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession Q969D9-1). 1 * 100 µG ACRBTSP-H52HB100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou