Recombinat Protein,Mouse TSLP Protein, His Tag,Mouse TSLP, His Tag (TSP-M52H8) is expressed from human 293 cells (HEK293). It contai ns AA Tyr 20 - Glu 140 (Accession Q9JIE6-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse TSLP Protein, His Tag,Mouse TSLP, His Tag (TSP-M52H8) is expressed from human 293 cells (HEK293). It contai ns AA Tyr 20 - Glu 140 (Accession Q9JIE6-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse TSLP Protein, His Tag,Mouse TSLP, His Tag (TSP-M52H8) is expressed from human 293 cells (HEK293). It contai ns AA Tyr 20 - Glu 140 (Accession Q9JIE6-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG ACRBTSP-M52H8100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou