Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at s 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at s 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at s 1 * 100 µG ACRBTTR-H5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou