Recombinat Protein,Human UCH-L1 Protein, His Tag,Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Me t 1 - Ala 223 (Accession NP004172).This protein carries a polyhistidine tag at the N-terminus. The protein has a cal 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human UCH-L1 Protein, His Tag,Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Me t 1 - Ala 223 (Accession NP004172).This protein carries a polyhistidine tag at the N-terminus. The protein has a cal 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human UCH-L1 Protein, His Tag,Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Me t 1 - Ala 223 (Accession NP004172).This protein carries a polyhistidine tag at the N-terminus. The protein has a cal 1 * 1 mg ACRBUC1-H5140-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou