Recombinat Protein,Human UCH-L1 Protein, His Tag,Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Me t 1 - Ala 223 (Accession NP004172).This protein carries a polyhistidine tag at the N-terminus. The protein has a cal 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human UCH-L1 Protein, His Tag,Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Me t 1 - Ala 223 (Accession NP004172).This protein carries a polyhistidine tag at the N-terminus. The protein has a cal 1 * 50 µG ACRBUC1-H5140-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou