Recombinat Protein,Human uPAR PLAUR Protein, His Tag,Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession AAH02788).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human uPAR PLAUR Protein, His Tag,Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession AAH02788).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human uPAR PLAUR Protein, His Tag,Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession AAH02788).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 200 µG ACRBUPR-H5226200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou