Recombinat Protein,Human Vasorin VASN Protein, His Tag,Human Vasorin, His Tag (VAN-H52H3) is expressed from human 293 cells (HEK293 ). It contains AA Cys 24 - Pro 575 (Accession AAH68575).This protein carries a polyhistidine tag at the C-terminus. T 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Vasorin VASN Protein, His Tag,Human Vasorin, His Tag (VAN-H52H3) is expressed from human 293 cells (HEK293 ). It contains AA Cys 24 - Pro 575 (Accession AAH68575).This protein carries a polyhistidine tag at the C-terminus. T 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Vasorin VASN Protein, His Tag,Human Vasorin, His Tag (VAN-H52H3) is expressed from human 293 cells (HEK293 ). It contains AA Cys 24 - Pro 575 (Accession AAH68575).This protein carries a polyhistidine tag at the C-terminus. T 1 * 1 mg ACRBVAN-H52H3-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou