Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession P15692-9) . 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession P15692-9) . 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession P15692-9) . 1 * 50 µG ACRBVE1-H5246-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou