Human VEGF165, His Tag (VE5-H5248) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession P15692-4 null
Fournisseur: ACROBIOSYSTEMS
Human VEGF165, His Tag (VE5-H5248) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession P15692-4 null
COMMANDE
Développer tout / Réduire tout
Human VEGF165, His Tag (VE5-H5248) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession P15692-4 null ACRBVE5-H5248-50UG Each
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou