MABSol Biotinylated Human VEGF165, epitope tag free, primary amine labeling (VE5-H8210) is expressed from human HEK293 cells. It c ontains AA Ala 27 - Arg 191 (Accession NP001165097). It is the biotinylated form of ActiveMax Human VEGF165, Tag Fr 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Human VEGF165, epitope tag free, primary amine labeling (VE5-H8210) is expressed from human HEK293 cells. It c ontains AA Ala 27 - Arg 191 (Accession NP001165097). It is the biotinylated form of ActiveMax Human VEGF165, Tag Fr 1 * 200 G
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human VEGF165, epitope tag free, primary amine labeling (VE5-H8210) is expressed from human HEK293 cells. It c ontains AA Ala 27 - Arg 191 (Accession NP001165097). It is the biotinylated form of ActiveMax® Human VEGF165, Tag Fr 1 * 200 µG ACRBVE5-H8210200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou