Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBVS2-H5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou