Recombinat Protein,Human VSIG4 Protein, His Tag,Human VSIG4, His Tag (VS4-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Arg 20 - Pro 283 (Accession AAH10525).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human VSIG4 Protein, His Tag,Human VSIG4, His Tag (VS4-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Arg 20 - Pro 283 (Accession AAH10525).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBVS4-H5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou