Recombinat Protein,Human VSIG8 Protein, His Tag,Human VSIG8, His Tag (VS8-H52H1) is expressed from human 293 cells (HEK293). It cont ains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human VSIG8 Protein, His Tag,Human VSIG8, His Tag (VS8-H52H1) is expressed from human 293 cells (HEK293). It cont ains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human VSIG8 Protein, His Tag,Human VSIG8, His Tag (VS8-H52H1) is expressed from human 293 cells (HEK293). It cont ains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG ACRBVS8-H52H1100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou