Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) GP, His Tag (ZE5-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Asp 637 (Accession AAQ55048.1). 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Ebolavirus EBOV (subtype Zaire, strain Kikwit-95) GP, His Tag (ZE5-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Asp 637 (Accession AAQ55048.1). 1 * 100 µG ACRBZE5-V5221100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou