Synonyms:
Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8
320780075
ACRO320780075EA
331
EUR
InStock
ACRO320780075
Tétrahydrofuranne-D8 99.5 atom % D pour spectroscopie RMN
Tétrahydrofuranne-D8
Packaged in 0.75 ml ampoules