Synonyms:
CHIR 99021, 6-((2-((4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)amino)nicotinonitrile, CHEMBL412142, GSK-3 Inhibitor XVI, AGN-PC-0MWAEX
468131000
ACRO468131000EA
409
EUR
InStock
ACRO468131000
CHIR99021 95%
CHIR99021
Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.
Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies