Surgical scissors, single use, Aesculap® SUSI®
Fournisseur: AESCULAP AG & CO
BC324SU
AESCBC324SUEA 124 EUR InStock
AESCBC324SU
Surgical scissors, single use, Aesculap® SUSI®
Ciseaux
These disposable scissors are made from a high performance polymer. The special composition of this high-grade polymer ensures an outstanding quality and provides optimal mechanical properties.
  • Single use reduces risk of cross-contamination
  • Reduction of processing costs in hospitals
  • Sterilised in a validated process
The material ensures a safe, easy and environmentally friendly removal through incineration in the regular hospital waste removal system.
Conditionnement: The scissors are sterile packed in single peel packs. The single peel packs are presented in dispenser boxes containing an indicated quantity of units as described in this leaflet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité