EletroTen-blue electroporation-competent cells
Fournisseur: AGILENT
200159 200159
STRA200159EA 424 EUR InStock
STRA200159 AGLS200159
EletroTen-blue electroporation-competent cells
Cellules compétentes
Achieve the highest transformation efficiencies available when cloning large plasmids and ligated DNA.
  • Separate DNA fragments and libraries up to 5000 bp in 1 to 2 min/sample
  • Achieve objective quantitative and purity measurements from as little as 2 µl of DNA, sensitive down to 5 pg/µl
  • Ideal for sample QC in NGS workflows
  • Partially used ScreenTape devices can be saved and used at a later time
  • No need to batch samples
Agilent EletroTen-Blue Electroporation-Competent Cells offer the highest transformation efficiencies available, dramatically improving cloning large plasmids and ligated DNA. These cells also offer the highest electroporation efficiency (Hee) phenotype, which confers increased resilience in harsh electroporation conditions.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité