ABLE C competent cells
Fournisseur: AGILENT
200172
AGLS200172EA 399 EUR InStock
AGLS200172
ABLE C competent cells
Cellules compétentes
ABLE Competent Cells for toxic clones reduce copy number four to ten fold compared to the XL1-Blue strain.
  • Reduces the copy number of ColE1-derived plasmids (such as pUC and pBluescript® plasmids) four-fold
  • Decreases level of cloned protein product to increase cell viability
  • Transformation efficiency of 5×10⁶ to 2×10⁷ transformants/µg
  • Also available in an electroporation-competent version
ABLE C Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE C strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
The ABLE C Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE C strains.
ABLE K Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE K strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
ABLE K Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE K strains.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité